Would you like to log in again?
Feilchenfeld Splinter Forceps, OR Grade, Sklar
Supplier: Sklar
19-2945 19-2930
10193-934EA 50.13 USD InStock
10193-934 10194-858
Feilchenfeld Splinter Forceps, OR Grade, Sklar
Forceps
Feilchenfeld Splinter Forceps are multi-purpose thumb forceps with sharp tips used, for removing small foreign objects.
They feature a wide thumb grip (central groove) on the handle for increased control and precision.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty