Would you like to log in again?
Cannon Curette, OR Grade, Sklar
Supplier: Sklar
06-4005 06-4007 06-4006 06-4004 06-4002 06-4003 06-4001
10194-868EA 251.25 USD InStock
10194-868 10194-274 10194-252 10194-870 10194-872 10194-874 10194-698
Cannon Curette, OR Grade, Sklar
Curettes
Sklar's Cannon Curette is a dermatological instrument used for scraping and removing superficial tissue or minor skin growths.
It is a stylus shaped tool with a fenestrated, oval tip that features sharp edges for cutting.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty