Would you like to log in again?
Dermal Curette Oval, OR Grade, Sklar
Supplier: Sklar
REVIEWS (0)
Total Ratings: 0
Avg. Ratings: out of 5
Sort all by:
06-4113 06-4115 06-4112 06-4114
10194-888EA 132.65 USD InStock
10194-888 10194-890 10194-892 10194-894
Dermal Curette Oval, OR Grade, Sklar
Curettes
Sklar's oval dermal curettes are used for scraping skin lesions and growths, such as warts or melanomas.
Censitrac Ready products are industry standard and constructed of stainless steel.
These are single-ended instruments with a loop that is sharp on the inside but blunt on the outside, so as not to damage the surrounding skin.
Ordering information: Censitrac instruments are custom barcoded to reflect the facility. This is unique customization. The initial order for the facility can take 14 to 30 days, subsequent orders can take less time.
Caution: Customers must have a Censitrac Site ID to purchase Censitrac instruments.
Order Now
ORDER
SPECIFICATIONS
WRITE A REVIEW
REVIEWS (0)
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty