Would you like to log in again?
Fox Dermal Curette, OR Grade, Sklar
Supplier: Sklar
REVIEWS (0)
Total Ratings: 0
Avg. Ratings: out of 5
Sort all by:
06-4139 06-4134 06-4141 06-4135 06-4140 06-4136 06-4137 06-4133 06-4138
10194-910EA 123.31 USD InStock
10194-910 10194-418 10194-192 10194-912 10194-914 10194-916 10194-918 10194-420 10194-920
Fox Dermal Curette, OR Grade, Sklar
Curettes
Sklar's Fox Dermal Curette is used for scraping skin lesions and growths such as warts or melanomas.
  • Premium OR-grade
  • Fenestrated working surface
  • Non sterile and latex-free
This single ended instrument is available with an oval or round loop tip. The loop is sharp on the inside and blunt on the outside so the lesion can be removed, but surrounding skin is not damaged. A variety of tip sizes are also available to accommodate varying lesion sizes. The handle of the Fox curette is flat and lightweight.
Order Now
ORDER
SPECIFICATIONS
WRITE A REVIEW
REVIEWS (0)
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty