Would you like to log in again?
Long Stitch Scissors, OR Grade, Sklar
Supplier: Sklar
15-1358
10195-004EA 227.99 USD InStock
10195-004
Long Stitch Scissors, OR Grade, Sklar
Scissors
Sklar's Long Stitch Scissors are used for suture removal
These scissors have a small hook-shaped tip on one blade that slides under sutures to slightly lift them before cutting for removal. This hook also holds the suture so it does not slip off the blade of the scissors prior to cutting. Long Stitch Scissors are commonly used for cutting sutures in gynecological procedures and oral surgery. They should not be used for cutting tissue. This device is straight with smooth, blunt/blunt tips.
Ordering information: Censitrac instruments are custom barcoded to reflect the facility. This is unique customization. The initial order for the facility can take 14 to 30 days, subsequent orders can take less time.
Caution: Customers must have a Censitrac Site ID to purchase Censitrac instruments.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty