Would you like to log in again?
Sklarcut™ Metzenbaum Dissecting Scissors, OR Grade, Sklar
Supplier: Sklar
Sklarcut™
15-3586 15-3574
10195-014EA 312.84 USD InStock
10195-014 10015-664
Sklarcut™ Metzenbaum Dissecting Scissors, OR Grade, Sklar
Scissors
Sklarcut™ Metzenbaum Dissecting Scissors are used for dissecting and cutting tissue only
Sklarcut™ scissors feature micro-serrations on one blade to reduce trauma when cutting both delicate and course tissue. These serrations assist in holding the tissue during cutting and minimizes tissue from slipping so the most accurate and clean cuts can be made. Sklarcut™ scissors are identified by a single gold coated finger ring on the handle of the serrated blade.
These scissors are not recommended for cutting sutures or drains.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty