Would you like to log in again?
New Orleans Needle Holder, OR Grade, Sklar
Supplier: Sklar
20-1870
10195-044EA 211.93 USD InStock
10195-044
New Orleans Needle Holder, OR Grade, Sklar
Forceps
New Orleans needle holder is a ratcheted, finger ring instrument designed for holding and controling needles during suturing.
The tip of this instrument is short, cross-serrated, and features a single center groove which provides a secure hold on the needle.
Censitrac Ready products are industry standard.
Ordering information: Censitrac instruments are custom barcoded to reflect the facility. This is unique customization. The initial order for the facility can take 14 to 30 days, subsequent orders can take less time.
Caution: Customers must have a Censitrac Site ID to purchase Censitrac instruments.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty