Would you like to log in again?
TC Debakey Cardiovascular Needle Holder, OR Grade, Sklar
Supplier: Sklar
REVIEWS (0)
Total Ratings: 0
Avg. Ratings: out of 5
Sort all by:
21-8092 21-8087 21-8090 21-8089 21-8088
10195-058EA 534.94 USD InStock
10195-058 10195-056 10195-054 10195-052 10195-194
TC Debakey Cardiovascular Needle Holder, OR Grade, Sklar
Forceps
Sklar's DeBakey Tungsten Carbide Cardiovascular Needle Holder with golden handle is designed for holding needles and suturing in cardiovascular procedures.
  • Stay sharp for a longer period of time
  • Tungsten carbide inserts provides maximizes control and precision
Censitrac Ready products are industry standard and constructed of stainless steel.
Ordering information: Censitrac instruments are custom barcoded to reflect the facility. This is unique customization. The initial order for the facility can take 14 to 30 days, subsequent orders can take less time.
Caution: Customers must have a Censitrac Site ID to purchase Censitrac instruments.
Order Now
ORDER
WRITE A REVIEW
REVIEWS (0)
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty