Would you like to log in again?
Econo™ Olsen-Hegar Needle Holder, Floor Grade, Sklar
Supplier: Sklar
21-877
10195-060BX 166.98 USD InStock
10195-060
Econo™ Olsen-Hegar Needle Holder, Floor Grade, Sklar
Forceps
The Econo™ Olsen-Hegar Needle Holder is a ratcheted, finger ring instrument that also has built-in scissors.
The Econo™ Olsen-Hegar Needle Holder is used with intermediate weight sutures, commonly in plastic surgery procedures. Econo™ instruments are floor grade, disposable instruments sold in bulk quantities.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty