Would you like to log in again?
Sklarlite XD™ Metzenbaum-Lahey Dissecting Scissors, OR Grade, Sklar
Supplier: Sklar
23-1178 23-1179 23-1177
10195-104EA 235.78 USD InStock
10195-104 10195-102 10195-106
Sklarlite XD™ Metzenbaum-Lahey Dissecting Scissors, OR Grade, Sklar
Scissors
Sklarlite XD™ Metzenbaum-Lahey Dissecting Scissors are most commonly used for dissecting and cutting tissue
These scissors are not recommended for cutting sutures, drains, or heavy tissue. Metzenbaum-Lahey scissors tend to be shorter than traditional Metzenbaum scissors. Sklarlite XD™ instruments are noticeably lighter and delicate than standard instruments which allow the surgeon the highest degree of precision with the least amount of fatigue. These instruments have been uniquely manufactured to be one of the lightest, most delicate general instruments available today.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty