Would you like to log in again?
Sklarlite XD™ Metzenbaum Dissecting Scissors, OR Grade, Sklar
Supplier: Sklar
REVIEWS (0)
Total Ratings: 0
Avg. Ratings: out of 5
Sort all by:
Sklarlite XD™
23-1186 23-1184 23-1176
10195-114EA 316.76 USD InStock
10195-114 10195-112 10195-100
Sklarlite XD™ Metzenbaum Dissecting Scissors, OR Grade, Sklar
Scissors
Sklarlite XD™ Metzenbaum Dissecting Scissors are used for dissecting and cutting tissue only
Sklarlite XD™ instruments are lighter and more delicate than standard instruments which allow the surgeon high precision with the least amount of fatigue. These instruments have been manufactured to be one of the lightest, most delicate general instruments available today.
These scissors are not recommended for cutting sutures, drains, or heavy tissue.
Order Now
ORDER
WRITE A REVIEW
REVIEWS (0)
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty