Would you like to log in again?
Sklarlite XD™ Lister Bandage Scissors, OR Grade, Sklar
Supplier: Sklar
Sklarlite XD™
23-1202 23-1203
10195-122EA 263.57 USD InStock
10195-122 10195-120
Sklarlite XD™ Lister Bandage Scissors, OR Grade, Sklar
Scissors
Sklarlite XD™ Lister Bandage Scissors are a versatile instrument with multiple uses
The jaws of the Lister Bandage Scissors are angled with the lower blade being slightly longer. The tip of the lower blade features a flattened blunt nodule that is intended to slide between bandages and skin without harming the skin. This feature allows Lister Bandage Scissors to occasionally be used during cesarean section to open the uterus without causing inadvertent injury to the baby. Ring handles can be equal in size or with one ring larger.
Sklarlite XD™ instruments are lighter and more delicate than standard instruments, which allow the surgeon the highest degree of precision with the least amount of fatigue.
They are used for sizing dressings and removing circumferential bandages. They may also be used to cut tougher material such as plaster, fabric and occasionally umbilical cord.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty