Would you like to log in again?
Scheduled maintenance of the site is planned from June 8th at 7:00 PM to June 9th at 5:00 PM ET. During this time, contract pricing and registration functionality will be unavailable. We appreciate your patience.
Sklarlite XD™ Derf Needle Holders, OR Grade, Sklar®
Supplier: Sklar
23-2224
10195-130EA 380.47 USD InStock
10195-130
Sklarlite XD™ Derf Needle Holders, OR Grade, Sklar®
Forceps
Sklarlite XD™ Derf Needle Holders feature short jaws and delicate serrations that hold small needles and commonly 5-0 and 6-0 sutures
Frequently utilized in ophthalmic, dental, or plastic procedures, the shorter length allows the Derf Needle Holder to be easily controlled in smaller surgical areas. Sklarlite XD™ instruments are noticeably lighter and delicate than standard instruments which allow the surgeon the highest degree of precision with the least amount of fatigue. These instruments have been uniquely manufactured to be one of the lightest, most delicate general instruments available today. This product is straight with serrated tips, a central groove and a length of 4-3/4 inches.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty