Would you like to log in again?
Sklarlite XD™ Mayo-Hegar Needle Holder, OR Grade, Sklar
Supplier: Sklar
23-2228
10195-134EA 228.51 USD InStock
10195-134
Sklarlite XD™ Mayo-Hegar Needle Holder, OR Grade, Sklar
Forceps
The Sklarlite XD™ Mayo-Hegar Needle Holder is a ratcheted, finger ring instrument available in a wide range of lengths for multiple types of surgical procedures
Due to it's versatility, it is one of the most commonly used surgical tools for both human and animal procdures. Shorter length products may be used in oral, gynecological, or dermatological procedures, as well as with small animals. Larger lengths of this product may be used for deep tissue suturing such as in bariatric procedures, or in large animal procedures. The Mayo-Hegar needle holder is commonly used with larger, heavier suture needles because of it's wide jaws. Sklarlite XD™ instruments are noticeably lighter and more delicate than standard instruments which allow the surgeon the highest degree of precision with the least amount of fatigue.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty