Would you like to log in again?
Sklarlite XD™ Crile-Wood Needle Holder, OR Grade, Sklar
Supplier: Sklar
23-2234
10195-138EA 357.9 USD InStock
10195-138
Sklarlite XD™ Crile-Wood Needle Holder, OR Grade, Sklar
Forceps
The Sklarlite XD™ Crile-Wood Needle Holder is used to hold and guide small to medium size needles and suture materials
It has a gently tapered, blunt tip and is generally more delicate than the Mayo-Hegar but slightly less delicate than Webster, Halsey, or DeBakey needle holders. The ratcheted finger ring handle aids in securing needles for both continuous and interrupted closures. Sklarlite XD™ instruments are noticeably lighter and delicate than standard instruments, providing surgeons with highest degree of precision and the least amount of fatigue. These instruments have been uniquely manufactured to be one of the lightest, most delicate general instruments available today.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty