Would you like to log in again?
Sklarlite XD™ Webster Needle Holder, OR Grade, Sklar
Supplier: Sklar
23-2220 23-2225 23-2235
10195-132EA 310.94 USD InStock
10195-132 10195-140 10195-128
Sklarlite XD™ Webster Needle Holder, OR Grade, Sklar
Forceps
Sklarlite XD™ Webster Needle Holders are used with small needles and fine suture material, commonly in plastic surgery or subcutaneous procedures
They are available with smooth or serrated jaws. Smooth jaws will cause the least damage to the needle, however serrated jaws will hold the needle more tightly. Physician preference typically determines the type of working surface utilized. Sklarlite XD™ instruments are noticeably lighter and more delicate than standard instruments, allowing the surgeon the highest degree of precision with the least amount of fatigue. These instruments have been uniquely manufactured to be one of the lightest, most delicate general instruments available today.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty