Would you like to log in again?
Sklarlite XD™ Kelly Hemostatic Forceps, OR Grade, Sklar
Supplier: Sklar
Sklarlite XD™
23-2507
10195-146EA 156.13 USD InStock
10195-146
Sklarlite XD™ Kelly Hemostatic Forceps, OR Grade, Sklar
Forceps
Sklarlite XD™ Kelly Hemostatic Forceps are primarily used for clamping large blood vessels or manipulating heavy tissue
Similar to Rankin forceps, the jaws of Kelly Forceps are 1/3 the length of the shanks and serrations are 1/2 the length of the jaws, however, Kelly forceps are shorter and heavier than Rankin forceps.
Sklarlite XD™ instruments are lighter and more delicate than standard instruments, which allow the surgeon the highest degree of precision with the least amount of fatigue.
They may be used for soft tissue dissection.
Ordering information: Available curved or straight with a ratcheted finger ring handle.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty