Would you like to log in again?
Sklarlite XD™ Crile Hemostatic Forceps, OR Grade, Sklar
Supplier: Sklar
Sklarlite XD™
23-2513 23-2510
10195-148EA 175.72 USD InStock
10195-148 10195-150
Sklarlite XD™ Crile Hemostatic Forceps, OR Grade, Sklar
Forceps
Sklarlite XD™ Crile Hemostatic Forceps, also known as a Crile Hemostat, are used for clamping blood vessels or tissue before cauterization or ligation.
Horizontal serrations the entire length of the jaw, and jaws half the length of the shank make this instrument unique. They may also be used for soft tissue dissection no deeper than 6-8" or in Laparotomy procedures. Sklarlite XD™ instruments are lighter and more delicate than standard instruments which allow the surgeon the highest degree of precision with the least amount of fatigue.
Ordering information: Censitrac instruments are custom barcoded to reflect the facility. This is unique customization. The initial order for the facility can take 14 to 30 days, subsequent orders can take less time.
Caution: Customers must have a Censitrac Site ID to purchase Censitrac instruments.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty