Would you like to log in again?
Scheduled maintenance of the site is planned from June 8th at 7:00 PM to June 9th at 5:00 PM ET. During this time, contract pricing and registration functionality will be unavailable. We appreciate your patience.
Sklarlite™ Mixter Right Angle Forceps, OR Grade, Sklar
Supplier: Sklar
Sklarlite™
23-2520 23-2516
10195-154EA 338.07 USD InStock
10195-154 10195-152
Sklarlite™ Mixter Right Angle Forceps, OR Grade, Sklar
Forceps
Sklarlite™ Mixter Right Angle Forceps are a lighter version of standard Mixter forceps for clamping vessels, dissection, or grasping tissue is obscured surgical areas.
This lightweight modification provides the surgeon with greater control and less fatigue, while the right angle of the jaws provides access to difficult sites. These forceps are a ratcheted, finger ring instrument that has serrations the entire length of the jaw and are able to provide a strong grip.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty