Would you like to log in again?
Sklarlite XD™ Mixter Right Angle Forceps, OR Grade, Sklar
Supplier: Sklar
Sklarlite XD™
23-2521 23-2525
10195-158EA 302.01 USD InStock
10195-158 10195-156
Sklarlite XD™ Mixter Right Angle Forceps, OR Grade, Sklar
Forceps
Sklarlite XD™ Mixter Right Angle Forceps are a lighter version of standard Mixter forceps for clamping vessels, dissection, or grasping tissue is obscured surgical areas
These forceps are a ratcheted, finger ring instrument, that has serrations the entire length of the jaw and are able to provide a strong grip.
This lightweight modification provides the surgeon with greater control and less fatigue, while the right angle of the jaws provides access to difficult sites.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty