Would you like to log in again?
Finochietto Needle Holder, OR Grade, Sklar
Supplier: Sklar
20-4510
10195-178EA 428.92 USD InStock
10195-178
Finochietto Needle Holder, OR Grade, Sklar
Forceps
Finochietto Needle Holder is a ratcheted, finger ring instrument most commonly used for suturing in OB/GYN procedures. The long shanks of this instrument make it ideal for suturing in deep, narrow areas. They are also used for colorectal anastomosis.
This product has straight shanks and curved, cross-serrated tips with a central groove.
Censitrac Ready products are industry standard and constructed of stainless steel.
Ordering information: Censitrac instruments are custom barcoded to reflect the facility. This is unique customization. The initial order for the facility can take 14 to 30 days, subsequent orders can take less time.
Caution: Customers must have a Censitrac Site ID to purchase Censitrac instruments.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty