Would you like to log in again?
TC Crile-Wood Left Handed Needle Holder, OR Grade, Sklar
Supplier: Sklar
21-9035 21-9030
10195-198EA 429.02 USD InStock
10195-198 10195-196
TC Crile-Wood Left Handed Needle Holder, OR Grade, Sklar
Forceps
Sklar's TC Crile-Wood Left-Handed Needle Holder is a instrument specially designed for left-handed users to hold and guide small to medium size needles and suture materials
The Crile-Wood Needle Holder has a gently tapered, blunt tip and is generally more delicate than the Mayo-Hegar but slightly less delicate than Webster, Halsey, or DeBakey needle holders. The ratcheted finger ring handle aids in securing needles for both continuous and interrupted closures. Tungsten Carbide instruments are designed to stay sharp for a significantly longer period of time than standard instruments. Sklar permanently bonds tungsten carbide inserts to the instrument, which maximizes control and precision.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty