Would you like to log in again?
Littler Suture Carrying Scissors, OR Grade, Sklar
Supplier: Sklar
22-2645
10195-212EA 175.65 USD InStock
10195-212
Littler Suture Carrying Scissors, OR Grade, Sklar
Scissors
Sklar's Littler Suture Carrying Scissors are finger ring scissors that feature a single tiny hole in each blade through which suture material is passed and carried around the surgical site
The blunt/blunt tips may also be used for dissecting fine tissue. This product is curved with smooth edges.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty