Would you like to log in again?
Sklarlite™ Sklarcut™ Operating Scissors, OR Grade, Sklar®
Supplier: Sklar
REVIEWS (0)
Total Ratings: 0
Avg. Ratings: out of 5
Sort all by:
23-1260 23-1262 23-1266 23-1264
10195-284EA 236.63 USD InStock
10195-284 76192-944 10195-282 10195-280
Sklarlite™ Sklarcut™ Operating Scissors, OR Grade, Sklar®
Scissors
Sklarlite™ Sklarcut™ Operating scissors, also known as Surgical scissors, are a versatile surgical instrument
  • For more than 120 years, Sklar has set the standard for surgical instrumentation
  • With five major surgical instruments lines, Sklar is able to deliver superior precision for any surgical procedure
  • All Sklar products are industry standard
Sklarlite™ Sklarcut™ scissors are a lighter version of the standard scissors, that feature micro-serrations on one blade to assist in securely holding the tissue during cutting and minimize tissue from slipping so the most accurate and atraumatic cuts can be made. A unique manufacturing process developed by Sklar shaves microns off standard pattern instruments while maintaining structural integrity and durability. This lightweight modification provides the surgeon with greater control and less fatigue.
Sklarcut™ scissors are identified by a single gold coated finger ring on the handle of the serrated blade.
With three available tip configurations, varying curvatures and multiple lengths, these scissors are used for general, multipurpose cutting and dissecting. Sharp Operating Scissors cut tougher tissue and are frequently used for dissecting, whereas blunt Operating Scissors are used for cutting materials such as sutures, tubing or gauze.
Order Now
ORDER
WRITE A REVIEW
REVIEWS (0)
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty