Would you like to log in again?
Sklarlite™ Sklarcut™ Mayo Dissecting Scissors, OR Grade, Sklar
Supplier: Sklar
Sklarlite™ Sklarcut™
23-1274 23-1278
10195-288EA 320.72 USD InStock
10195-288 10195-290
Sklarlite™ Sklarcut™ Mayo Dissecting Scissors, OR Grade, Sklar
Scissors
Sklarlite™ Sklarcut™ Mayo Dissecting Scissors are a lighter version of the standard Mayo scissors, that also feature ultra-fine serrations on one blade
While the curved instrument is used for cutting or dissecting dense or deep tissue, and the straight version is used for cutting surface tissue or sutures, both may be used interchangeably depending on procedure and physician preference. Sklarlite™ Sklarcut™ scissors are identified by a single gold coated finger ring for the serrated blade.
The lightweight modification provides the surgeon with greater control and less fatigue, while the micro-serrations reduce trauma as well as hold the tissue during cutting. Mayo Scissors are a versatile instrument that can be used in multiple procedures and settings.
Ordering information: Available straight or curved in a variety of lengths with a standard beveled blade.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty