Would you like to log in again?
Sklarlite™ Derf Needle Holder, OR Grade, Sklar
Supplier: Sklar
Sklarlite™
23-2124
10195-320EA 316.29 USD InStock
10195-320
Sklarlite™ Derf Needle Holder, OR Grade, Sklar
Forceps
Sklarlite™ Derf Needle Holders are used in ophthalmic, dental, or plastic procedures
They feature short jaws and delicate serrations, that hold small needles and 5-0 and 6-0 sutures.
Sklarlite™ instruments are a lighter version of standard Sklar instruments. A manufacturing process developed by Sklar shaves microns off standard and delicate pattern instruments while maintaining structural integrity and durability. This lightweight modification provides the surgeon with greater control and less fatigue.
The shorter length allows the Derf Needle Holder to be controlled in smaller surgical areas.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty