Would you like to log in again?
Scheduled maintenance of the site is planned from June 8th at 7:00 PM to June 9th at 5:00 PM ET. During this time, contract pricing and registration functionality will be unavailable. We appreciate your patience.
Sklarlite™ Halsey Needle Holder, OR Grade, Sklar
Supplier: Sklar
Sklarlite™
23-2130
10195-326EA 360.14 USD InStock
10195-326
Sklarlite™ Halsey Needle Holder, OR Grade, Sklar
Forceps
vSklarlite™ Halsey Needle Holders are used for holding small needles for suturing, during plastic surgery and procedures with delicate tissue, such as dermatology.
Smooth jaws will cause the least damage to the needle; however, serrated jaws hold the needle more tightly. Physician preference typically determines the type of working surface utilized.
Sklarlite™ instruments are a lighter version of standard Sklar instruments. A manufacturing process developed by Sklar shaves microns off standard and delicate pattern instruments, while maintaining structural integrity and durability. This lightweight modification provides the surgeon with greater control and less fatigue.
Ordering information: Available with smooth or delicately serrated jaws.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty