Would you like to log in again?
Sklarlite™ Crile-Wood Needle Holder, OR Grade, Sklar
Supplier: Sklar
Sklarlite™
23-2134
10195-330EA 297.52 USD InStock
10195-330
Sklarlite™ Crile-Wood Needle Holder, OR Grade, Sklar
Forceps
The Sklarlite™ Crile-Wood Needle Holder is used to hold and guide small to medium size needles and suture materials.
The Crile-Wood Needle Holder has a gently tapered, blunt tip and is generally more delicate than the Mayo-Hegar but slightly less delicate than Webster, Halsey, or DeBakey needle holders. The ratcheted finger ring handle aids in securing needles for both continuous and interrupted closures.
Sklarlite™ instruments are a lighter version of standard Sklar instruments. A manufacturing process developed by Sklar shaves microns off standard and delicate pattern instruments while maintaining structural integrity and durability. This lightweight modification provides the surgeon with greater control and less fatigue.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty