Would you like to log in again?
Sklarlite™ Allis Tissue Forceps, OR Grade, Sklar
Supplier: Sklar
23-2861 23-2851 23-2862 23-2852 23-2864 23-2865
10195-378EA 247.52 USD InStock
10195-378 10195-376 10195-374 10195-372 10195-370 10195-380
Sklarlite™ Allis Tissue Forceps, OR Grade, Sklar
Forceps
Sklarlite™ Allis Tissue Forceps are finger ring, ratcheted forceps used for grasping organs and slippery or dense tissue during electrosurgery
Sklarlite™ instruments are a lighter version of standard Sklar instruments. A unique manufacturing process developed by Sklar shaves microns off standard and delicate pattern instruments while maintaining structural integrity and durability. This lightweight modification provides the surgeon with greater control and less fatigue.
They are used with tonsil, vaginal, breast, and thyroid tissue.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty