Would you like to log in again?
Sklarlite™ Littauer Cutting Forceps, OR Grade, Sklar
Supplier: Sklar
REVIEWS (0)
Total Ratings: 0
Avg. Ratings: out of 5
Sort all by:
23-9276 23-9273 23-9251 23-9255 23-9259 23-9275
10195-388EA 223.18 USD InStock
10195-388 10195-386 10195-384 10195-382 10195-392 10195-390
Sklarlite™ Littauer Cutting Forceps, OR Grade, Sklar
Bone Cutters and Rongeurs
Sklarlite™ Littauer Cutting Forceps are a lighter version of the standard, non-ratcheted, plier handle instrument for cutting nail and bone
The short, sharp blades are able to produce enough force to cut through thick nail and small bone, but delicate enough not to cause trauma to surrounding tissue or organs. This is helpful with nail debridement which should be completed in short, small sections to avoid damage to the hyponychium.
This lightweight modification provides the user with greater control and less fatigue. As nail nippers they are used for nail debridement. They may also be used to nip delicate bones found in the facial and nasal region, or small animal (mice) veterinary procedures.
Order Now
ORDER
WRITE A REVIEW
REVIEWS (0)
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty