Would you like to log in again?
Michel Applying and Removing Forceps, OR Grade, Sklar
Supplier: Sklar
24-1250
10195-402EA 241.41 USD InStock
10195-402
Michel Applying and Removing Forceps, OR Grade, Sklar
Forceps
Michel Applying and Removing Forceps are thumb forceps, with a single spring mechanism, used to apply and remove surgical clips.
Clips may be applied to wounds and removed when healing is complete or they may be used as part of a procedure to stop blood flow.
This instrument has serrated edges, so the clip can be gripped for proper placement and secure removal.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty