Would you like to log in again?
Kirschner Wire, OR Grade, Sklar
Supplier: Sklar
REVIEWS (0)
Total Ratings: 0
Avg. Ratings: out of 5
Sort all by:
40-1535 40-1565 40-1566 40-1580 40-1567 40-1545 40-1582 40-1562 40-1572 40-1584 40-1571 40-1569 40-1568
10195-808EA 12.89 USD InStock
10195-808 10194-330 10193-908 10192-356 10192-284 10193-836 10193-736 10192-364 10193-834 10192-270 10193-744 10195-514 10185-280
Kirschner Wire, OR Grade, Sklar
Trocars
Kirschner Wire, also known as K-Wire, is used to stabilize internal bone fragments in both human and veterinary procedures
They are available in several different diameters and lengths, as well as smooth or threaded. Threaded K-Wires, although more difficult to place and remove, tend to provide a more secure hold and do not migrate to unintended areas of the body. K-Wire is inserted directly through the skin with the assistance of a guidance system, and is removed once the bone has healed.
Order Now
ORDER
WRITE A REVIEW
REVIEWS (0)
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty