Would you like to log in again?
Steinmann Pin Trocar Point, OR Grade, Sklar
Supplier: Sklar
REVIEWS (0)
Total Ratings: 0
Avg. Ratings: out of 5
Sort all by:
40-1592 40-1590 40-1591 40-1595 40-1594 40-1593 40-1596
10195-522EA 31.14 USD InStock
10195-522 10195-520 10195-518 10195-516 10195-526 10193-896 10195-524
Steinmann Pin Trocar Point, OR Grade, Sklar
These are thin metal rods with trocar point commonly used in orthopedic procedures for holding large bone fractures together.
  • Made of stainless steel
These pins function similarly to K-Wire, but are larger in diameter.
Order Now
ORDER
WRITE A REVIEW
REVIEWS (0)
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty