Would you like to log in again?
Mead Rongeur, OR Grade, Sklar®
Supplier: Sklar
REVIEWS (0)
Total Ratings: 0
Avg. Ratings: out of 5
Sort all by:
40-3208
10195-640EA 610.03 USD InStock
10195-640
Mead Rongeur, OR Grade, Sklar®
Bone Cutters and Rongeurs
Mead spring handle rongeur is used in orthopedic procedures for handling or cutting small bones. The curved jaw beaks ensures a firm grip on the bone and eases gouging areas that may have otherwise been harder to reach.
Order Now
ORDER
SPECIFICATIONS
WRITE A REVIEW
REVIEWS (0)
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty