Would you like to log in again?
Singley (Tuttle) Forceps, OR Grade, Sklar
Supplier: Sklar
36-1290
10195-724EA 228.26 USD InStock
10195-724
Singley (Tuttle) Forceps, OR Grade, Sklar
Forceps
Singley (Tuttle) Forceps are thumb forceps used for grasping intestinal tissue or dressings materials and sponges
This product is straight with oval, fenestrated and serrated tips.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty