Would you like to log in again?
Adair Breast Tenaculum Forceps, OR Grade, Sklar®
Supplier: Sklar
36-1865
10195-730EA 261.32 USD InStock
10195-730
Adair Breast Tenaculum Forceps, OR Grade, Sklar®
Forceps
Sklar® Adair Breast Tenaculum Forceps are a ratcheted, finger ring instrument used to penetrate and grasp fibrocystic tissue or masses
They are perforating grasping forceps with sharp, non-crushing prong tips. This product has straight shanks with inward curving, single prong tips.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty