Would you like to log in again?
Rochester-Pean Hemostatic Forceps, OR Grade, Sklar
Supplier: Sklar
REVIEWS (0)
Total Ratings: 0
Avg. Ratings: out of 5
Sort all by:
22-3555 36-2790 22-3655 36-2690 36-2810 36-2710 22-2280
10195-754EA 250.13 USD InStock
10195-754 10195-752 10193-702 10195-750 89025-280 89025-360 89025-362
Rochester-Pean Hemostatic Forceps, OR Grade, Sklar
Forceps
Rochester-Pean Hemostatic Forceps are used for clamping larger tissue and vessels for hemostasis.
The full horizontal serrations and availability in multiple lengths make these forceps a versatile instrument used in multiple procedures
Censitrac Ready OR Grade Rochester-Pean Forceps are industry standard and constructed of stainless steel.
Available curved or straight.
Ordering information: Censitrac instruments are custom barcoded to reflect the facility. This is unique customization. The initial order for the facility can take 14 to 30 days, subsequent orders can take less time.
Caution: Customers must have a Censitrac Site ID to purchase Censitrac instruments.
Order Now
ORDER
DOCUMENTATION
WRITE A REVIEW
REVIEWS (0)
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty