Would you like to log in again?
Allen Intestinal Forceps, OR Grade, Sklar
Supplier: Sklar
REVIEWS (0)
Total Ratings: 0
Avg. Ratings: out of 5
Sort all by:
36-3180 36-3195
10195-764EA 364.89 USD InStock
10195-764 10195-762
Allen Intestinal Forceps, OR Grade, Sklar
Forceps
Censitrac Ready products are industry standard and constructed of stainless steel.
  • Sklar's® Allen Intestinal Forceps are a ratcheted, finger ring clamp primarily used for grasping tissue or hemostasis. It is very similar to a toothed Kocher clamp, but is more lightweight.
Ordering information: Censitrac instruments are custom barcoded to reflect the facility. This is unique customization. The initial order for the facility can take 14 to 30 days, subsequent orders can take less time.
Caution: Customers must have a Censitrac Site ID to purchase Censitrac instruments.
Order Now
ORDER
WRITE A REVIEW
REVIEWS (0)
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty