Would you like to log in again?
Leksell Rongeur, OR Grade, Sklar®
Supplier: Sklar
40-4097 40-4095 40-4092 40-4098
10195-830EA 1555.65 USD InStock
10195-830 10195-836 10195-834 10195-832
Leksell Rongeur, OR Grade, Sklar®
Bone Cutters and Rongeurs
Leksell Rongeur is a double action, plier handle instrument used for debulking, primarily in orthopedic procedures, but occasionally neurosurgical operations as well.
  • Available angled or with a slight curve
  • Features a scooping bite which removes bone and tissue from the spinal area
Order Now
ORDER
SPECIFICATIONS
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty