Would you like to log in again?
Mallet with Two Replaceable Flat Plastic Ends, OR Grade, Sklar
Supplier: Sklar
40-5926 40-5925
10195-938PK 108.12 USD InStock
10195-938 10195-936
Mallet with Two Replaceable Flat Plastic Ends, OR Grade, Sklar
Mallets and Hammers
Mallet with Two Replaceable Flat Plastic Ends are constructed of stainless steel and plastic
Mallet heads are 2¼" wide with a 25mm diameter. The overall weight is 0.48lbs. Industry Standard.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty