Would you like to log in again?
Cobb Type Spinal Gouge, OR Grade, Sklar
Supplier: Sklar
REVIEWS (0)
Total Ratings: 0
Avg. Ratings: out of 5
Sort all by:
40-6921 40-6920 40-6923 40-6922
10196-164EA 442.81 USD InStock
10196-164 75817-948 10196-162 75817-950
Cobb Type Spinal Gouge, OR Grade, Sklar
Gouges
These non sterile cobb type spinal gouges are designed for carving out bone or making holes in close spaces of the vertebral canal.
  • Round knurled handle
  • Reusable, latex-free
  • 5/32" blade width
  • Industry standard
Order Now
ORDER
WRITE A REVIEW
REVIEWS (0)
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty