Would you like to log in again?
Williger Elevator, OR Grade, Sklar®
Supplier: Sklar
40-7155
10196-204EA 99.6 USD InStock
10196-204
Williger Elevator, OR Grade, Sklar®
Retractors
Sklar's® Williger Elevator is an instrument used in bone surgery to lift periosteum from bones or dissect soft tissue.
  • The elevator is used to along the bone but does not cut into it
This product features a round, tapered tip so it does not damage surrounding tissue.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty