Would you like to log in again?
Scheduled maintenance of the site is planned from June 8th at 7:00 PM to June 9th at 5:00 PM ET. During this time, contract pricing and registration functionality will be unavailable. We appreciate your patience.
DeBakey Atraumatic Tissue Forceps, OR Grade, Sklar
Supplier: Sklar
52-6587 52-6589 52-6654 52-6588 52-6957
10196-464EA 669.05 USD InStock
10196-464 10196-466 10196-462 10196-248 10197-094
DeBakey Atraumatic Tissue Forceps, OR Grade, Sklar
Forceps
DeBakey Atraumatic Pediatric Multipurpose Clamp is a ratcheted, finger ring instrument, that may be used as a hemostat to prevent blood flow, or as grasping forceps since the atraumatic teeth provide a secure grip but do not harm the tissues.
The multipurpose clamp is angled 30, 60 or 90 degrees.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty