Would you like to log in again?
DeBakey Bainbridge Cardiovascular Forceps, OR Grade, Sklar
Supplier: Sklar
52-6702
10196-274EA 481.55 USD InStock
10196-274
DeBakey Bainbridge Cardiovascular Forceps, OR Grade, Sklar
Forceps
Sklar's DeBakey Bainbridge Atraumatic Forceps are a ratcheted, finger ring instrument commonly used in cardiovascular procedures
It's ratcheted locking mechanism allows this instrument to be used as a hemostat to control blood flow. The atraumatic teeth also allow it to be used for grasping fine or delicate tissue with little to no damage to the tissue itself. This product is straight with longitudinally serrated tips, 1x2 rows of fine, atraumatic teeth.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty