Would you like to log in again?
DeBakey Peripheral Vessel Clamp, OR Grade, Sklar®
Supplier: Sklar
52-6603
10196-480EA 666.74 USD InStock
10196-480
DeBakey Peripheral Vessel Clamp, OR Grade, Sklar®
Forceps
Sklar's® DeBakey Atraumatic Peripheral Vascular Clamp is a ratcheted, finger ring clamp used in cardiovascular procedures for clamping and halting blood flow from larger vessels
This product has a double/'S' curve with 1x2 rows of fine, atraumatic teeth, and a length of 8 inches.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty