Would you like to log in again?
DeBakey Atraumatic Peripheral Vessel Clamp, OR Grade, Sklar
Supplier: Sklar
52-6605 52-6600 52-6604
10196-482EA 656.33 USD InStock
10196-482 10196-474 10196-484
DeBakey Atraumatic Peripheral Vessel Clamp, OR Grade, Sklar
Forceps
DeBakey Atraumatic Peripheral Vascular Clamp is a ratcheted, finger ring clamp used in cardiovascular procedures for clamping and halting blood flow from larger vessels
Sklar operating room grade instruments are made from the highest grade surgical steel and incorporate the finest level of craftsmanship. Superior manufacturing and quality inspection guarantee that Sklar instruments meet customer demands. These instruments have been designed to provide exceptional comfort and precision all while ensuring the durability and strength that has been provided by Sklar for over 100 years.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty