Would you like to log in again?
McGlamry Elevator, OR Grade, Sklar
Supplier: Sklar
REVIEWS (0)
Total Ratings: 0
Avg. Ratings: out of 5
Sort all by:
40-7480 40-7484 40-7482 40-7486
10196-498EA 236.58 USD InStock
10196-498 10196-500 10196-502 10196-504
McGlamry Elevator, OR Grade, Sklar
Retractors
McGlamry Elevators are constructed of stainless steel.
  • Industry standard
These elevators used to deglove a metatarsal head and free fused sesamoidal plates.
Order Now
ORDER
WRITE A REVIEW
REVIEWS (0)
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty