Would you like to log in again?
Hayes Hand and Finger Retractor, OR Grade, Sklar
Supplier: Sklar
40-8265
10196-556EA 119.51 USD InStock
10196-556
Hayes Hand and Finger Retractor, OR Grade, Sklar
Retractors
Hayes Hand and Finger Retractors feature a 11/2" blade length, that tapers from 3/8" to 1/8" in width
Constructed of stainless steel. Industry standard.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty