Would you like to log in again?
Adson Retractor, OR Grade, Sklar®
Supplier: Sklar
42-1785 42-1775 42-1511 42-1510 42-1611
10196-684EA 1073.98 USD InStock
10196-684 10196-692 10196-680 10196-682 10196-690
Adson Retractor, OR Grade, Sklar®
Retractors
Adson Retractor is a finger ring, self-retaining retractor used during intracranial procedures to hold back the cerebellum
It is available with sharp or blunt interlocking prongs in various configurations, angled or straight arms, and multiple lengths.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty