Would you like to log in again?
Joseph Nasal Knife, OR Grade, Sklar®
Supplier: Sklar
41-1260 41-1266 41-1274 41-1270 41-1262 41-1276 41-1264
10196-434EA 185.78 USD InStock
10196-434 10196-704 10196-706 10196-700 10196-436 10196-702 10196-438
Joseph Nasal Knife, OR Grade, Sklar®
Knives
Sklar's® Joseph Knife is an ENT/plastic surgery instrument frequently used in nasal reconstructive procedures
After the Joseph button end knife is used to separate the nasal mucosa from the nasal cartilage, the double-edged Joseph knife is used to shave away targeted cartilage based on the procedure. This instrument is available straight or curved and with a sharp or blunt tip, determined by the needs of the surgeon.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty